Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Parainfluenza Virus Type 3 Monoclonal Antibody (H103A), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA17316
Description
The MA1-7316 antibody reacts with Parainfluenza virus type 3 in viral and infected tissue samples. MA1-7316 has been successfully used in ELISA and Immunofluorescence applications. The MA1-7316 antibody was raised against Parainfluenza Type 3 antigen.
Specifications
Parainfluenza Virus Type 3 | |
Monoclonal | |
0.1 mg/mL | |
hPIV-3; PIV-3; Parainfluenza Virus Type 3 | |
Parainfluenza Type 3 antigen. | |
100 μg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunofluorescence | |
H103A | |
Unconjugated | |
Mouse | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title