Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pararosaniline base, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
467-62-9
C19H19N3O
305.38
MFCD00036222
KRVRUAYUNOQMOV-UHFFFAOYSA-N
pararosaniline base, tris 4-aminophenyl methanol, tris p-aminophenyl methanol, benzenemethanol, 4-amino-.alpha.,.alpha.-bis 4-aminophenyl, tris 4-aminophenyl methan-1-ol, fuchsin dye base, parafuchsin carbinol, tris 4-aminophenyl carbinol, methanol, tris 4-aminophenyl, 4,4',4-triaminotrityl alcohol
10084
tris(4-aminophenyl)methanol
C1=CC(=CC=C1C(C2=CC=C(C=C2)N)(C3=CC=C(C=C3)N)O)N
467-62-9
C19H19N3O
305.38
MFCD00036222
KRVRUAYUNOQMOV-UHFFFAOYSA-N
pararosaniline base, tris 4-aminophenyl methanol, tris p-aminophenyl methanol, benzenemethanol, 4-amino-.alpha.,.alpha.-bis 4-aminophenyl, tris 4-aminophenyl methan-1-ol, fuchsin dye base, parafuchsin carbinol, tris 4-aminophenyl carbinol, methanol, tris 4-aminophenyl, 4,4',4-triaminotrityl alcohol
10084
tris(4-aminophenyl)methanol
C1=CC(=CC=C1C(C2=CC=C(C=C2)N)(C3=CC=C(C=C3)N)O)N
Melting Point 205.0°C
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 50g
Color Purple
Beilstein 13, 733
Solubility Information Solubility in water: very slightly soluble. Other solubilities: soluble in alcohol,slightly soluble in ether
Formula Weight 305.38
Physical Form Crystalline Powder
Chemical Name or Material Pararosaniline base

GHS H Statement
Suspected of causing cancer.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.

GHS Signal Word: Warning

EINECSNumber : 207-395-