Alfa Aesar™ Pararosaniline hydrochloride, 90+%

pH indicator 1.2 (purple) to 3.0 (red)

Manufacturer: Alfa Aesar™ 3654030

Quantity 250g

 View more versions of this product

Catalog No. AA3654030

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 569-61-9
    Formula Weight 323.83
    MDL Number MFCD00001657
    Beilstein 4164603
    Synonym Basic Red 9 monohydrochloride; C.I. 42500
    Chemical Name or Material Pararosaniline hydrochloride

    Specifications

    Formula Weight 323.83
    Health Hazard 3 GHS P Statement
    P281-P201-P202-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Obtain special instructions before use.
    Do not handle until all safety precautions have been read and understood.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Beilstein 4164603
    Health Hazard 2 GHS H Statement
    H350
    May cause cancer.
    Melting Point 268° to 270°C (dec.)
    RTECS Number CX9850100
    Physical Form Powder
    Quantity 250g
    MDL Number MFCD00001657
    CAS 569-61-9
    Synonym Basic Red 9 monohydrochloride; C.I. 42500
    Health Hazard 1 Danger
    TSCA TSCA
    Chemical Name or Material Pararosaniline hydrochloride
    Assay ≥90%

    GHS H Statement
    H350
    May cause cancer.

    GHS P Statement
    P281-P201-P202-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Obtain special instructions before use.
    Do not handle until all safety precautions have been read and understood.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit