Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Parathyroid Hormone (44-68), human, Alfa Aesar™

$142.40 - $239.50

Specifications

Format Solid
CAS 64421-69-8
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ66604EXD View Documents Alfa Aesar
J66604EXD
0.5mg Each for $142.40
Order in Bulk

Request bulk or custom formats

Add to cart
 
AAJ66604MCR View Documents Alfa Aesar
J66604MCR
1mg Each for $239.50
Order in Bulk

Request bulk or custom formats

Add to cart
 
Specifications

Specifications

Solid
64421-69-8
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit