Learn More
PARD3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA549856
Catalog No. PA549856
Add to cart
Description
Description
The antibody detects endogenous levels of total PARD3 protein.
PARD proteins, which were first identified in C. elegans, are essential for asymmetric cell division and polarized growth, whereas CDC42 mediates the establishment of cell polarity. The CDC42 GTPase, which is controlled by nucleotide exchange factors and GTPase-activating proteins domain.
Specifications
Specifications
PARD3
Polyclonal
Unconjugated
PARD3
Liquid
PARD3
Synthesized peptide derived from internal of human PARD3.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q8TEW0, Q99NH2, Q9Z340
PAR3, PAR3A
Rabbit
IgG
100 μL
-20°C
Primary
56288, 81918, 93742