Learn More
PARG Mouse anti-Human, Clone: OTI1B5, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF808508
Catalog No. 50-166-8897
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Poly(ADP-ribose) glycohydrolase (PARG) is the major enzyme responsible for the catabolism of poly(ADP-ribose), a reversible covalent-modifier of chromosomal proteins. The protein is found in many tissues and may be subject to proteolysis generating smaller, active products.
Specifications
Specifications
PARG
Monoclonal
Unconjugated
PARG
Lyophilized
PARG
Human recombit protein fragment corresponding to amino acids 1-367 of human PARG produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
OTI1B5
PBS with 8% trehalose and no preservative; pH 7.3
Q86W56
PARG99
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
8505