Learn More
Parker Balston Nitrogen Generators: N2-14 Series from Restek™
Safe, reliable, and requires low maintenance
$737.00 - $15650.00
Specifications
Certifications/Compliance CE
For Use With (Application) Turns compressed air into ultra-pure nitrogen
View More Specs
Products 2
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
06-710-898
Accessories View Documents Promotion Details
Restek CorporationSupplier Diversity Partner
20677
Nitrogen Generator N2-14
Each for $15,650.00
Add to cart
06-711-702
View Documents Promotion Details
Restek CorporationSupplier Diversity Partner
21648
Maintenance Kit for Models N2-14, N2-14A, 75-72, 75-720NA
Each for $737.00
Add to cart
Description
Description
  • Turns compressed air into ultra-pure nitrogen (up to 99.5%)
  • Model N2-14 requires no electricity
Specifications
Specifications
CE
Turns compressed air into ultra-pure nitrogen