Learn More
Parker domnick hunter™ Demicap™ Filter Capsules, Peplyn Plus Membrane Available on GSA/VA Contract for Federal Government customers only.
Compact, cost-effective, and efficient method of filtration for small-scale applications and batch filtration
Manufacturer:  Parker domnick hunter™ FEPPA1.0MMPN3
 View more versions of this product
Catalog No. 14-950-308
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
1μm
No
USP Plastics Class VI (121°C) certified, ASTM F838-83 certified
7.87 in.
0.3m2
1/4 in. NPT Male
Polypropylene
Yes
For clarification, sterilization and prefiltration applications
200mm
1/4 in. NPT Male