Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Parker domnick hunter™ Demicap™ Filter Capsules, Peplyn Plus Membrane - 5.5 in. (140mm) Length

Compact, cost-effective, and efficient method of filtration for small-scale applications and batch filtration

Manufacturer:  Parker domnick hunter™ FEPPB1.0GGPN3

 View more versions of this product

Catalog No. 14-950-327


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

1μm
No
USP Plastics Class VI (121°C) certified, ASTM F838-83 certified
5.5 in.
0.15m2
Stepped Hosebarb
Polypropylene
Yes
For clarification, sterilization and prefiltration applications
140mm
Stepped Hosebarb
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit