Learn More
Parker domnick hunter™ Demicap™ Filter Capsules, Prepor GF Membrane - 0.6µm Pore Size Available on GSA/VA Contract for Federal Government customers only.
Compact, cost-effective, and efficient method of filtration for small-scale applications and batch filtration
Manufacturer:  Parker domnick hunter™ FEGFA.60GGPN3
 View more versions of this product
Catalog No. 14-950-363
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
6μm
No
USP Plastics Class VI (121°C) certified, ASTM F838-83 certified
7.87 in.
0.22m2
Propor GF Pleated Glass Microfiber
Yes
For clarification, sterilization and prefiltration applications
200mm
Stepped Hosebarb