Learn More
Parker domnick hunter™ Demicap™ Filter Capsules, Prepor PES Membrane - 0.2µm Pore Size Non-distribution product as customer accommodation.
Compact, cost-effective, and efficient method of filtration for small-scale applications and batch filtration
Manufacturer:  Parker domnick hunter™ FEPSB020HHPS3
 View more versions of this product
Catalog No. 14-950-265
Please call customer service for current pricing. 1-800-766-7000
Description
Description
Capsule design minimizes hold-up volumes while providing high flowrates without the need for cartridge housings.
  • Particulate rated polyethersulfone membrane with 0.1 to 1.20 micron ratings
  • H.A. area for prefiltration life
  • Good chemical compatibility and low extractables
  • Maximum operating conditions, in direction of flow (outside to in): 72.5psi at 104°F for liquid filtration; and 58psi for air/gas
  • Capsules fully validated by 107 organisms/cm2 minimum bacterial challenge
  • Autoclavable by steam sterilization
  • Available in three sizes with a choice of inlet and outlet connections including Tri-Clamp, threaded, and hosebarbs
USP Plastics Class VI (121°C) certified, ASTM F838-83 certified
Applications:
For clarification, sterilization and prefiltration applications
Specifications
Specifications
Yes
0.12m2
1/2 in. Hosebarb
140mm
0.2μm
USP Plastics Class VI (121°C) certified, ASTM F838-83 certified
For clarification, sterilization and prefiltration applications
5.5 in.
Polyethersulfone
Sterile