Learn More
Parker domnick hunter™ Demicap™ Filter Capsules, Propor PES Membrane - 0.1micrometer Pore Size, Polypropylene Available on GSA/VA Contract for Federal Government customers only.
Compact, cost-effective, and efficient method of filtration for small-scale applications and batch filtration
Manufacturer:  Parker domnick hunter™ FEMSB010HHPSX
 View more versions of this product
Catalog No. 14-950-146
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
0.1μm
Yes
USP Plastics Class VI (121°C) certified, ASTM F838-83 certified
5.5 in.
0.22m2
Polyethersulfone; Polypropylene
Yes
For clarification, sterilization and prefiltration applications
140mm
1/2 in. Hosebarb