Learn More
Parker domnick hunter™ Demicap™ Filter Capsules, Propor PES Membrane - 0.2micrometer Pore Size, Nylon Non-distribution product as customer accommodation.
Low chemical and protein binding characteristics
Manufacturer:  Parker domnick hunter™ FEMSA020HHPS3
 View more versions of this product
Catalog No. 14-950-119
Add to cart
Description
Description
Compact, cost-effective, and efficient method of filtration for small-scale applications and batch filtration. Capsule design minimizes hold-up volumes while providing high flowrates without the need for cartridge housings.
  • Polyethersulfone membrane
  • Naturally hydrophilic and easy to integrity test
  • Maximum operating conditions, in direction of flow (outside to in): 72.5psi at 104°F for liquid filtration; and 58psi for air/gas
  • Capsules fully validated by 107 organisms/cm2 minimum bacterial challenge
  • Autoclavable by steam sterilization
  • Available in three sizes with a choice of inlet and outlet connections including Tri-Clamp, threaded, and hosebarbs
Certifications
ASTM F838-83 certified
Specifications
Specifications
1/2 in. Hosebarb
200 mm
USP Plastics Class VI (121°C) certified, ASTM F838-83 certified
0.2 μm
PES, Nylon
7.87 in.
Autoclavable
Sterile
0.22 m2