Learn More
Parker domnick hunter™ Demicap™ Filter Capsules, Propor PES Membrane - 0.2micrometer Pore Size, Nylon Available on GSA/VA Contract for Federal Government customers only.
Low chemical and protein binding characteristics
Manufacturer:  Parker domnick hunter™ FEMSA020GGPS3
 View more versions of this product
Catalog No. 14-950-120
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
0.2μm
Yes
USP Plastics Class VI (121°C) certified, ASTM F838-83 certified
200mm
Stepped Hosebarb
Polyethersulfone; Nylon
Yes
7.87 in.
0.22m2