Learn More
Parker domnick hunter™ HIGH FLOW TETPOR II Air/Gas Filters Available on GSA/VA Contract for Federal Government customers only.
Provide full retention of bacteria, viruses, and particulate in process air and gas streams, with optimum efficiency, flow rate, and strength, and zero fiber shedding
Manufacturer:  Parker domnick hunter™ ZHFT/1C
 View more versions of this product
Catalog No. 14-950-420
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Conforms to current USP Plastics Class VI - 121°C, ASTM F838-83
C Style
0.2μm
Lyophilizers, autoclave vacuum breaks and blow fill seal equipment
25.4cm
0.2μm
Silicone
Stainless Steel
Locking 226
Sterile gas applications, including fermentation inlet and off gas stream, particle free air applications in the electronics industry
10 in.
PTFE
High Flow Tetpor Ii Air/Gas Filter