Learn More
Parker domnick hunter™ Service Kit for Pure Air Gas Generators
Manufacturer:  Parker domnick hunter™ 8000 DK2
Catalog No. 14-950-17
Add to cart
Description
Description
Includes replacement parts necessary for annual (8000-hour) service of domnick hunter™ pure air gas generators. Consists of one replacement for each of two inlet filter elements, one outlet filter element, one drain, and one moisture indicator kit.
Specifications
Specifications
Service Kit
1 replacement for two inlet filter elements (1), outlet filter element (1), drain (1), moisture indicator kit (1)
Annual (8000 hours) service of pure air gas generators