Parker domnick hunter™ Service Kits for Nitrogen Gas Generators
Manufacturer: Parker domnick hunter™ 606260630
 View more versions of this product
Catalog No. 14-950-23
  Check Availability
Add to Cart
    Specifications
    Product Type Service Kit
    For Use With (Equipment) Semi annual (4000 hours) service, uhplcms nitrogen gas generator
    Includes Two each of exhaust silencers, compressor inlet filter felts, sintered inserts, 4 mist-x silencers
  • Description & Specifications
Related Products
Product Content Feedback