Learn More
Parker domnick hunter™ Stackable Zero-Air Generators Available on GSA/VA Contract for Federal Government customers only.
Modular and stackable with domnick hunter hydrogen generators
Manufacturer:  Parker domnick hunter™ UHP150ZASE
 View more versions of this product
Catalog No. 14-950-609
  Check Availability
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Add to cart
Specifications Description & Specifications
Specifications
Zero Air Generator
CE, UL
42.5cm
45.5cm
5° to 40°C
15.2kg
230V 50/60Hz
58 to 145psi
2 Years
16.7 in.
17.9 in.
50/60Hz
230V
13.4 in.
UHP-150ZA-S
575w