Learn More
Parker domnick hunter™ Zero-Air Generators Non-distribution product as customer accommodation.
Generate an uninterrupted supply of clean zero-purity air using in-house low-pressure compressed air supply.
Manufacturer:  Parker domnick hunter™ UHP35ZA
 View more versions of this product
Catalog No. 14-950-13
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Alert:
Filter elements must be changed once a year or when indicator/gauge changes to red.
Description
Description
Ideal for GC-FID, emissions analyzers and other gas chromatography applications using NPD and FPD that require zero air free from oil, water and particulates. Ensure consistent flat GC baselines and reduce cleaning requirements of FIDs.
  • Three-stage bench- or wall-mounted generators use a 1μm coalescing filter with Oil-X™Plus technology to remove oil, water and particulates. A heated platinum catalyst converts hydrocarbons, including methane, to carbon dioxide and water before a final pass through a high-efficiency Oil-XPlus 0.01μm particulate filter
  • Total hydrocarbon concentration of <0.1ppm (as methane) is achievable with inlet hydrocarbon concentration less than 10ppm (methane)
  • Operates over the temperature range 35° to 150°F
  • The noiseless unit features an autodrain on the inlet filter A green plastic pop-up indicator on each filter housing indicates when the increase in differential pressure caused by oil and dirt contamination requires the filter to be changed
  • Inlet/Outlet Connection: 1/4 in. Swagelok™
†230V 50/60Hz models also available. For more information, contact your Fisher Scientific™ Customer Service Representative.
Compliance
CE, CSA, UL compliant.
Warranty and Services
Die-cast filter housing with inner and outer Alocrom corrosion protection has 10-year warranty.
Specifications
Specifications
3500 cc/min.
SDS
Documents
Product Certifications
Accessories