Parker domnick hunter™ Nitrogen Generators for Analytical Instruments (ICP)
Manufacturer: Parker domnick hunter™ HPN25000E
 View more versions of this product
Catalog No. 14-950-630
  Check Availability
Add to Cart
    Specifications
    Product Type Ultra High Purity Nitrogen Generators
    Electrical Requirements 207-253V 50/60Hz
    Includes Without integral compressor
    Nitrogen Purity >99.999%
    Flow Rate 5L/min
    For Use With (Application) Analytical Instruments (ICP)
  • Description & Specifications
Related Products
Product Content Feedback