Learn More
Parker domnick hunter™ Nitrogen Generators for Analytical Instruments (ICP)
Manufacturer:  Parker domnick hunter™ HPN25000W
 View more versions of this product
Catalog No. 14-950-631
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Analytical Instruments (ICP)
5L/min
Without integral compressor
Ultra High Purity Nitrogen Generators
103 -126V 60Hz
>99.999%