Parker domnick hunter™ Nitrogen Generators for Analytical Instruments (ICP)
Manufacturer: Parker domnick hunter™ HPN25000CE
 View more versions of this product
Catalog No. 14-950-632
  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Product Type Ultra High Purity Nitrogen Generators
    Electrical Requirements 207-253V 50/60Hz
    Includes With integral compressor
    Nitrogen Purity >99.999%
    Flow Rate 5L/min
    For Use With (Application) Analytical Instruments (ICP)
  • Description & Specifications
Related Products
Product Content Feedback