Learn More
Parker domnick hunter™ Nitrogen Generators for GC
Manufacturer:  Parker domnick hunter™ UHPN2750E
 View more versions of this product
Catalog No. 14-950-614
  Check Availability
Add to cart
Specifications
Description
  • Produce ultra high purity nitrogen for critical life science, chemical analysis and spectroscopy applications
  • Continuous stream of ultra-high purity nitrogen from single ‘plug & play’ unit
  • Integral oil free compressor, with advanced noise reduction technology
  • Eliminates inconvenient and potentially dangerous nitrogen cylinders
  • Compact, reliable with minimal operator attention and maintenance
  • Flow rates range: 0.75L/min to 3L/min
  • Purity: >99.9995%
Description & Specifications
Specifications
GC and other critical analytical applications
0.75L/min
Without Integral Compressor
Ultra High Purity Nitrogen Generators
207-253V 50/60Hz
>99.9995%