Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Parker domnick hunter™ Nitrogen Generators for GC

Manufacturer:  Parker domnick hunter™ UHPN2750W

 View more versions of this product

Catalog No. 14-950-615


  Check Availability
Add to cart

Specifications

Description

  • Produce ultra high purity nitrogen for critical life science, chemical analysis and spectroscopy applications
  • Continuous stream of ultra-high purity nitrogen from single ‘plug & play’ unit
  • Integral oil free compressor, with advanced noise reduction technology
  • Eliminates inconvenient and potentially dangerous nitrogen cylinders
  • Compact, reliable with minimal operator attention and maintenance
  • Flow rates range: 0.75L/min to 3L/min
  • Purity: >99.9995%
Description & Specifications

Specifications

GC and other critical analytical applications
0.75L/min
Without Integral Compressor
Ultra High Purity Nitrogen Generators
103-126V 60Hz
>99.9995%
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit