Parker domnick hunter™ Nitrogen Generators for GC
Manufacturer: Parker domnick hunter™ UHPN23000CE
 View more versions of this product
Catalog No. 14-950-624
  Check Availability
Add to Cart
    Specifications
    Product Type Ultra High Purity Nitrogen Generators
    Electrical Requirements 207-253V 50/60Hz
    Includes With Integral Compressor
    Nitrogen Purity >99.9995%
    Flow Rate 3L/min
    For Use With (Application) GC and other critical analytical applications
  • Description & Specifications
Related Products
Product Content Feedback