Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Parkin Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP180515
Description
Parkin Polyclonal specifically detects Parkin in Mouse samples. It is validated for Western Blot.Specifications
Parkin | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
NP_057903 | |
PRKN | |
Synthetic peptide directed towards amino acids 311-360 of the mouse Park2. Peptide sequence YTRYQQYGAEECVLQMGGVLCPRPGCGAGLLPEQGQRKVTCEGGNGLGCG. | |
100 μL | |
Primary | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS & 2% Sucrose. with 0.09% Sodium Azide | |
E3 ubiquitin ligase, E3 ubiquitin-protein ligase parkin, EC 6.3.2.-, LPRS2, parkin, parkin 2, Parkinson disease protein 2, Parkinson juvenile disease protein 2, parkinson protein 2, E3 ubiquitin protein ligase (parkin), PDJjuvenile) 2, parkin | |
Rabbit | |
Affinity Purified | |
RUO | |
5071 | |
Human, Mouse, Rat, Porcine, Bovine, Canine | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title