Learn More
PARN Mouse anti-Human, Clone: OTI3G9, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF802652
Catalog No. 50-166-6080
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene is a 3 ft.-exoribonuclease, with similarity to the RNase D family of 3 ft.-exonucleases. It prefers poly(A) as the substrate, hence, efficiently degrades poly(A) tails of mRNAs. Exonucleolytic degradation of the poly(A) tail is often the first step in the decay of eukaryotic mRNAs. This protein is also involved in silencing of certain maternal mRNAs during oocyte maturation and early embryonic development, as well as in nonsense-mediated decay (NMD) of mRNAs that contain premature stop codons. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
PARN
Monoclonal
Unconjugated
PARN
Lyophilized
PARN
Human recombit protein fragment corresponding to amino acids 1-266 of human PARN produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI3G9
PBS with 8% trehalose and no preservative; pH 7.3
O95453
DAN, DKCB6, PFBMFT4
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
5073