Learn More
PARP-2 Polyclonal Antibody, Active Motif™
SDP
Rabbit Polyclonal Antibody
Supplier:  Active Motif 39044
Encompass
 View more versions of this product
Catalog No. 50-199-2819
Add to Cart
Description
Description
This gene encodes polytransferase, but lacks an N-terminal DNA binding domain which activates the C-terminal catalytic domain of poly transferase. The basic residues within the N-terminal region of this protein may bear potential DNA-binding properties, and may be involved in the nuclear and/or nucleolar targeting of the protein. Two alternatively spliced transcript variants encoding distinct isoforms have been found.
Specifications
Specifications
PARP-2
Polyclonal
Unconjugated
PARP2
ADP-ribosyltransferase (NAD+, poly(ADP-ribose) polymerase)-like 2; ADP-ribosyltransferase (NAD+; poly (ADP-ribose) polymerase) 2; ADP-ribosyltransferase (NAD+; poly(ADP-ribose) polymerase)-like 2; ADP-ribosyltransferase diphtheria toxin-like 2; ADPRT2; ADPRT-2; Adprtl2; ADPRTL3; ARTD2; Aspartl2; C78626; DNA ADP-ribosyltransferase PARP2; HGNC:272; hPARP-2; LOC100351185; mPARP-2; NAD(+) ADP-ribosyltransferase 2; pADPRT-2; PARP2; PARP-2; poly (ADP-ribose) polymerase 2; poly (ADP-ribose) polymerase family, member 2; poly (ADP-ribose) polymerase family, member 2-like; poly (ADP-ribosyl) transferase-like 2; Poly [ADP-ribose] polymerase 2; poly(ADP-ribose) polymerase 2; Poly[ADP-ribose] synthase 2; poly[ADP-ribose] synthetase 2; Protein poly-ADP-ribosyltransferase PARP2
Rabbit
Antigen affinity chromatography
RUO
10038
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ChIP Assay, Immunoprecipitation, Western Blot
Lot-Specific
70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
Q9UGN5
PARP2
This PARP-2 antibody was raised against a recombinant protein corresponding to the C-terminal region of human PARP-2.
50 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions