Learn More
Epredia™ Lab Vision™ PARP (Poly ADP-Ribose Polymerase) Ab-2 Rabbit Polyclonal Antibody with BSA and azide
Deliver accurate, reproducible results with the Epredia™ PARP (Poly ADP-Ribose Polymerase) Ab-2, Rabbit Polyclonal Antibody.
Specifications
Antigen | PARP (Poly ADP-Ribose Polymerase) Ab-2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
RB1516R1
|
Epredia™
RB1516R1 |
Prediluted | 1 mL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
Description
Poly ADP-Ribose Polymerase (PARP) uses nicotinamide adenine dinucleotide (oxidized form) NAD as a substrate to catalyse the transfer of ADP-ribose to a variety of nuclear protein acceptors. Proteolysis of PARP to its stable 85kDa fragment is an early marker of programmed cell death (apoptosis) and is mediated by the caspase CPP32 protein. Cleavage occurs between Adp216 and Gly217, a site in PARP conserved across species.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human, Mouse and Rat. Others not known.
Epitope: N-terminal
Immunogen: A synthetic peptide for the N-terminal region of human PARP
Molecular Weight: 116kDa
Positive Control: Raji cells, Tonsil
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin only)
- Western Blotting
Note:
USA: RUO; Int'l: RUO
Specifications
PARP (Poly ADP-Ribose Polymerase) Ab-2 | |
Polyclonal | |
Rabbit | |
Apoptosis | |
A synthetic peptide for the N-terminal region of human PARP | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
Human, Mouse | |
IgG |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.