Learn More
R&D Systems™ PARP Treated Protein Control for Western Blot
Poly [ADP-ribose] Polymerase Control
Manufacturer:  R&D Systems™ 450010P
Catalog No. 45-001-0P
Add to cart
This item is not returnable. View return policy
Description
Description
Store the unopened product at -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Specifications
Specifications
10 μL
Store the unopened product at -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.