Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PARP1 Mouse anti-Human, Clone: 5F1, Abnova™

Mouse monoclonal antibody raised against full length recombinant human PARP1.

Manufacturer:  Abnova Corporation MAB12896

Catalog No. 89-934-726


Add to cart

Description

Description

This gene encodes a chromatin-associated enzyme, poly(ADP-ribosyl)transferase, which modifies various nuclear proteins by poly(ADP-ribosyl)ation. The modification is dependent on DNA and is involved in the regulation of various important cellular processes such as differentiation, proliferation, and tumor transformation and also in the regulation of the molecular events involved in the recovery of cell from DNA damage. In addition, this enzyme may be the site of mutation in Fanconi anemia, and may participate in the pathophysiology of type I diabetes. [provided by RefSeq]

Specifications

Specifications

PARP1
5F1
Mouse monoclonal antibody raised against full length recombinant human PARP1.
In PBS, pH 7.3 (50% glycerol, 1% BSA, 0.02% sodium azide).
ADPRT/ADPRT1/PARP/PARP-1/PPOL/pADPRT-1
Mouse
IgG1
50 uL
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot (1:2000) The optimal working dilution should be determined by the end user.
Liquid
PARP1
Recombinant protein corresponding to full length human PARP1.
Immunoaffinity purification
RUO
Primary
142
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit