Learn More
PARP1 Monoclonal Antibody (HC2R8), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$229.00
Specifications
Antigen | PARP1 |
---|---|
Clone | HC2R8 |
Concentration | 0.5 mg/mL |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-135-09
|
Invitrogen
14666782 |
100 μg |
Each of 1 for $229.00
|
|
|||||
Description
Description: This HC2R8 monoclonal antibody reacts with human poly (ADP-ribose) polymerase (PARP1). This ubiquitous 116 kDa nuclear enzyme is involved in DNA repair. During apoptosis, active caspases -3, -6 and -7 cleave PARP1 after Asp214, thereby inactivating PARP1 and generating two apoptotic fragments sized 85 kDa and 25 kDa. The HC2R8 antibody specifically recognizes both the cleaved and non-cleaved forms of PARP. Applications Reported: This HC2R8 antibody has been reported for use in immunoblotting (WB). Applications Tested: This HC2R8 antibody has been tested by immunoblotting of staurosporine-treated Jurkat cells. This can be used at less than or equal to 0.1 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
Poly ADP-Ribose Polymerase (PARP) uses nicotinamide adenine dinucleotide (oxidized form) NAD as a substrate to catalyse the transfer of ADP-ribose to a variety of nuclear protein acceptors. Proteolysis of PARP to its stable 85kDa fragment is an early marker of programmed cell death (apoptosis) and is mediated by the caspase CPP32 protein. Cleavage occurs between Adp216 and Gly217, a site in PARP conserved across species.Specifications
PARP1 | |
0.5 mg/mL | |
Unconjugated | |
Mouse | |
Human | |
142 | |
Primary | |
4° C |
HC2R8 | |
Monoclonal | |
Liquid | |
RUO | |
P09874 | |
IgG2b κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.