Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PARP12 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5142209
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.
Poly(ADP-ribosyl)ation is an immediate DNA-damage-dependent post-translational modification of histones and other nuclear proteins that contributes to the survival of injured proliferating cells. Poly(ADP-ribose) polymerases (PARPs) now constitute a large family of 18 proteins, encoded by different genes and displaying a conserved catalytic domain in which PARP-1 (113 kDa), the founding member, and PARP-2 (62 kDa) are so far the sole enzymes whose catalytic activity has been shown to be immediately stimulated by DNA strand breaks. A large repertoire of sequences encoding novel PARPs now extends considerably the field of poly(ADP-ribosyl)ation reactions to various aspects of the cell biology including cell proliferation and cell death. Some of these new members interact with each other, share common partners and common subcellular localizations suggesting possible fine tuning in the regulation of this post-translational modification of proteins.
Specifications
PARP12 | |
Polyclonal | |
Unconjugated | |
PARP12 | |
9930021O16; AA409132; AA536654; ADP-ribosyltransferase diphtheria toxin-like 12; ARTD12; MST109; MSTP109; PARP12; PARP-12; poly (ADP-ribose) polymerase family, member 12; poly [ADP-ribose] polymerase 12; poly ADP-ribose polymerase 12; poly(ADP-ribose) polymerase family member 12; Protein mono-ADP-ribosyltransferase PARP12; ZC3H1; Zc3hdc1; Zinc finger CCCH domain-containing protein 1; zinc finger CCCH type domain containing 1 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
64761 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9H0J9 | |
PARP12 | |
Peptide with sequence C-NAHDIKNKSSAP. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction