Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PARP12 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Specifications
Antigen | PARP12 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB028367
|
Novus Biologicals
NBP293240100UL |
100 μg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
NB028368
|
Novus Biologicals
NBP29324020UL |
20 μg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
PARP12 Polyclonal antibody specifically detects PARP12 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
PARP12 | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
EC 2.4.2.30, FLJ22693MST109, PARP-12, poly (ADP-ribose) polymerase family, member 12, poly [ADP-ribose] polymerase 12, ZC3H1, ZC3HDC1MSTP109, Zinc finger CCCH domain-containing protein 1, zinc finger CCCH type domain containing 1, zinc finger CCCH-type domain containing 1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 462-701 of human PARP12 (NP_073587.1). | |
Primary | |
Store at -20°:C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
Apoptosis, DNA Repair, Hypoxia | |
PBS (pH 7.3), 50% glycerol | |
64761 | |
IgG | |
Affinity purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title