Learn More
PARP15 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 181261AP
Catalog No. 50-173-2000
Add to cart
Description
Description
Transcriptional repressor. Has ADP-ribosyltransferase activity.
Specifications
Specifications
PARP15
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q460N3
BAL3, PARP 15, PARP15
Rabbit
IgG
150 μL
-20°C
Primary
165631
Western Blot
Unconjugated
PARP15
Liquid
PARP15
PARP15 Fusion Protein Ag12702
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human