Learn More
PARP2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 205551AP
Catalog No. 50-173-2959
Add to cart
Description
Description
The antibody is specific to PARP2 (C-terminal).
This gene encodes polytransferase, but lacks an N-terminal DNA binding domain which activates the C-terminal catalytic domain of poly transferase. The basic residues within the N-terminal region of this protein may bear potential DNA-binding properties, and may be involved in the nuclear and/or nucleolar targeting of the protein. Two alternatively spliced transcript variants encoding distinct isoforms have been found.
Specifications
Specifications
PARP2
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O88554, Q9UGN5
ADPRT2, ADPRT-2, ADPRTL2, ARTD2, hPARP-2, pADPRT-2, PARP2, PARP-2, Poly [ADP-ribose] polymerase 2
Rabbit
IgG
150 μL
-20°C
Primary
10038, 11546
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PARP2
Liquid
PARP2
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse