Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PARP6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531988
Description
Recommended positive controls: Jurkat, BCL-1. Predicted reactivity: Mouse (100%), Rat (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Poly(ADP-ribosyl)ation is an immediate DNA-damage-dependent post-translational modification of histones and other nuclear proteins that contributes to the survival of injured proliferating cells. Poly(ADP-ribose) polymerases (PARPs) now constitute a large family of 18 proteins, encoded by different genes and displaying a conserved catalytic domain in which PARP-1 (113 kDa), the founding member, and PARP-2 (62 kDa) are so far the sole enzymes whose catalytic activity has been shown to be immediately stimulated by DNA strand breaks. A large repertoire of sequences encoding novel PARPs now extends considerably the field of poly(ADP-ribosyl)ation reactions to various aspects of the cell biology including cell proliferation and cell death. Some of these new members interact with each other, share common partners and common subcellular localizations suggesting possible fine tuning in the regulation of this post-translational modification of proteins.
Specifications
PARP6 | |
Polyclonal | |
Unconjugated | |
Parp6 | |
1700119G14Rik; 2310028P13Rik; 3110038K10Rik; ADP-ribosyltransferase diphtheria toxin-like 17; ARTD17; C030013N01Rik; PARP6; PARP-6; PARP-6-B1; PARP-6-C; pART17; poly (ADP-ribose) polymerase family, member 6; poly [ADP-ribose] polymerase 6; poly(ADP-ribose) polymerase family member 6; Protein mono-ADP-ribosyltransferase PARP6; RGD1310937 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
56965, 67287 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.79 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q2NL67, Q6P6P7 | |
Parp6 | |
Recombinant fragment corresponding to a region within amino acids 339 and 575 of Human PARP6. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction