Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PARP9 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548254
Description
PARP9 Polyclonal antibody specifically detects PARP9 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
PARP9 | |
Polyclonal | |
Unconjugated | |
PARP9 | |
ADP-ribosyltransferase diphtheria toxin-like 9; ARTD9; AW214463; b aggressive lymphoma protein; B aggressive lymphoma protein homolog; Bagl; Bal; BAL1; BC003281; MGC:7868; Parp9; PARP-9; poly (ADP-ribose) polymerase 9; poly (ADP-ribose) polymerase family, member 9; poly [ADP-ribose] polymerase 9; poly(ADP-ribose) polymerase family member 9; Protein mono-ADP-ribosyltransferase PARP9; RGD1307534 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
80285, 83666 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide | |
Q8CAS9, Q8IXQ6 | |
PARP9 | |
KLH conjugated synthetic peptide between 599-629 amino acids from the C-terminal region of human Parp9. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction