Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PARS2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548121

Catalog No. PA548121

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene encodes a putative member of the class II family of aminoacyl-tRNA synthetases. These enzymes play a critical role in protein biosynthesis by charging tRNAs with their cognate amino acids. This protein is encoded by the nuclear genome but is likely to be imported to the mitochondrion where it is thought to catalyze the ligation of proline to tRNA molecules. Mutations have been found in this gene in some patients with Alpers syndrome. [provided by RefSeq, Mar 2015]
Specifications

Specifications

PARS2
Polyclonal
PBS with 0.09% sodium azide
Q7L3T8, Q8CFI5
Probable proline--tRNA ligase, mitochondrial, Prolyl-tRNA synthetase, ProRS, PARS2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
230577, 25973
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PARS2
Liquid
PARS2
KLH conjugated synthetic peptide between 390-417 amino acids from the C-terminal region of human PARS2.
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit