Learn More
Parter Medical™ 24-Hour Urine Containers
Proprietary, leak-resistant closure system assures specimen integrity during collection and transport to the laboratory
Manufacturer:  Parter Medical™ 126925
 View more versions of this product
Catalog No. 22-150-119
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
70mm
Vertical space saver
24 Hour Urine Container
Yes
No
HDPE
Used by both the patients and the health care professional for collection and transport of specimen to the laboratory
Natural, 70mm leak resistant HDPE closure with gasketted seal
3.5L
Screw cap
Amber
40 Case