Parter Medical™ Midstream Kits
Manufacturer: Parter Medical™ 273016
 View more versions of this product
Catalog No. 22-150-337
  Check Availability
Add to Cart
    Specifications
    Product Type Specimen Container
    Closure Color Natural
    For Use With (Application) For collection and transport of microbiology, Urinalysis, pathology, drug testing and histology specimens
    Capacity 0.023 gal., 90mL
    Includes Funnel, I.D. label, Blue cap, three-pack towelettes and user instructions
    Capacity (English) 0.023 gal.
    Sterile Yes
    Capacity (Metric) 90mL
  • Description & Specifications
Related Products
Product Content Feedback