Learn More
Parter Medical Products™ Nonsterile Specimen Containers
Designed for collection and transport of microbiology, urinalysis, pathology, histology and drug test specimens
$397.50
Specifications
Sterility Non-sterile
View More Specs
Products 2
Catalog Number Mfr. No. Capacity (Metric) Color Label Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Color Label Price Quantity & Availability  
22-150-114
View Documents
Parter Medical Products™
143211
90 mL White No Label
Case of 400 for $397.50
Add to cart
22-150-339
View Documents
Parter Medical Products™
142016
120 mL Natural English This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
  • Straight-sided container format featuring top and bottom positive grip knurls for ease of opening and closing when wearing protective or latex gloves
  • Stackable, leak-resistant and graduated unless otherwise noted
  • Incorporate the proprietary PMP™ Sure-Seal™ integrated sealing system and the proprietary PMP Sure-Lok™ proof-of-closure system
  • Manufactured from virgin polypropylene (container) and virgin polyethylene (closure) without the use of plasticizers or demolding additives; all material used free from latex and bovine contamination
  • Manufactured under strict process control and lot numbers are fully traceable
  • 95kpa compliant; testing results available upon request
  • Rigorous testing at varying pressures without seal failure proves this container ideal for use in pneumatic tube systems and harsh transport conditions; leak performance data is available upon request
  • Available in individually wrapped (Flo-Pac™) formats
  • Closures secure at 10 to 12 in.-lb. closing force
  • Reagent or fixative pre-fills may require 15 to 17 in.-lb. closing force to prevent post production cap-back-off
Specifications
Specifications
Non-sterile