Parter Medical™ Nonsterile Specimen Containers
Designed for collection and transport of microbiology, urinalysis, pathology, histology and drug test specimens
Manufacturer: Parter Medical™ 145616
 View more versions of this product
Catalog No. 22-150-348
  Check Availability
Add to Cart
    Specifications
    Sterile No
    Color Yellow
    Label English
    Closure Color Yellow
    Graduated Yes
    Size (Opening) (Metric) 48mm
    Capacity (Metric) 60mL
  • Description & Specifications
Related Products
Product Content Feedback