Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Parthenolide, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
100mg
This item is not returnable. View return policy
20554-84-1
C15H20O3
248.322
KTEXNACQROZXEV-KIFYNCCSSA-N
--parthenolide
131673949
CC1=CCCC2(C(O2)C3C(CC1)C(=C)C(=O)O3)C
This item is not returnable. View return policy
20554-84-1
C15H20O3
248.322
KTEXNACQROZXEV-KIFYNCCSSA-N
--parthenolide
131673949
CC1=CCCC2(C(O2)C3C(CC1)C(=C)C(=O)O3)C

5-HT release inhibitor

The active principle of feverfew (Chrysanthemum parthenium). Antisecretory, anti-inflammatory and spasmolytic; inhibits the release of 5-HT from blood platelets. Also inhibits activation of NF-κB, and generation of leukotriene B4 and thromboxane B2.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 100mg
Formula Weight 248.32
Percent Purity >98%
Chemical Name or Material Parthenolide

Recommended Storage : Desiccate at +4°C