Learn More
3M™ Particulate Respirator, N95 Products for Science Education
Lightweight and comfortable with snug, secure seal.
Products 2
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
S47062A Products for Science Education
View Documents
3M™
7100132742/EMD
Standard
Pack of 20 for $42.34
Case of 8 PK for $176.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
S47062 Products for Science Education
View Documents
3M™
18999473 EMW
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
N/A
Description
Description
At least 95% NIOSH tested filter efficiency for use where solids or non-oil-based liquid aerosols are present.
  • Synthetic elastic headband
  • Adjustable nose clip helps provide a custom fit and reduces the potential for eyewear fogging
  • Soft nose foam
  • 3M Catalog# 8210
These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product.
Order Info
Sold in packs of 20.
Compliance
Meets NIOSH 42CFR84 N95 requirements; NIOSH approval TC-84A-0007.