Learn More
Parvalbumin Rabbit anti-Human,Mouse,Rat, Unconjugated, Clone: 4G3, MilliporeSigma™
Rabbit Monoclonal Antibody
Manufacturer:  MilliporeSigma ZRB121825UL
 View more versions of this product
Catalog No. ZRB121825MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Parvalbumin Monoclonal antibody specifically detects Parvalbumin in Human,Mouse,Rat samples. It is validated for Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blotting.
Specifications
Specifications
Parvalbumin
4G3
Unconjugated
Purified
PVALB
GST/His-tagged full-length recombinant human Parvalbumin.
Target Molecular Weight 12.06 kDa calculated; ∼12 kDa observed. Uncharacterized bands may be observed in some lysate(s).
25 μL
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Monoclonal
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
30 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
NP_002845.1
Parvalbumin alpha;PV
Rabbit
IgG
Protein A Purified
Neuroscience
Primary
Human, Mouse, Rat