Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Parvin gamma Mouse anti-Human, Clone: 8C5.2, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen Parvin gamma
Clone 8C5.2
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, Immunoprecipitation 1-2 ug per 100-500 ug of protein 1ml of lysate
Applications Western Blot, Immunoprecipitation
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB123144
View Documents
Novus Biologicals
NBP30783520UG
20 μg
Each for $138.37
Add to cart
 
NB123143
View Documents
Novus Biologicals
NBP30783510UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

Parvin gamma Monoclonal antibody specifically detects Parvin gamma in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications

Specifications

Parvin gamma
0.2 mg/ml
Western Blot, Immunoprecipitation
Purified
RUO
10 mM PBS with 0.05% BSA
64098
IgG1 Kappa
Primary
Store at 4°C.
8C5.2
Western Blot 1-2 ug/ml, Immunoprecipitation 1-2 ug per 100-500 ug of protein 1ml of lysate
Unconjugated
Mouse
Human
gamma-parvin, parvin, gamma
GST-tagged recombinant protein corresponding to human Parvin gamma
Monoclonal
Protein A or G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit