Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PASD1 Mouse anti-Human, Janelia Fluor 646, Clone: 2ALCC136, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP250235JF646

Catalog No. NBP50235646


Add to cart

Description

Description

PASD1 Monoclonal antibody specifically detects PASD1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

PASD1
2ALCC136
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, KnockDown Validated
Purified
PASD1
GST-PASD1 recombinant protein containing 195-474 aas of PASD1. Region is common to PASD1a and PASD1b proteins.
Protein G purified
Store at 4°C in the dark.
Monoclonal
PASD1-1 recognizes the recombinant protein (aa 195-474) present in both PASD1_v1 and PASD1_v2 proteins.
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, KnockDown
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
CT63Cancer/testis antigen 63, OXTES1, OX-TES-1, PAS domain containing 1, PAS domain-containing protein 1
Mouse
IgG
0.1mL
Primary
139135
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit