Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PASK Mouse anti-Human, Clone: OTI11D8, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA500878

Catalog No. 50-167-2022


Add to cart

Description

Description

PASKIN (PAS-Kinase) is thought to regulate protein synthesis in cells. It is a conserved gene product found in yeast as well as mammals. PASKIN contains two domains (PAS A and PAS B) as well as a serine/threonine kinase domain related to AMP kinases. PASKIN activity leads to decreased carbohydrate storage, as well as increased protein synthesis. PASKIN-dependent phosphorylation inhibits the activity of mammalian glycogen synthase.
Specifications

Specifications

PASK
Monoclonal
0.79 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q96RG2
PASKIN, STK37
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
23178
Immunocytochemistry, Immunofluorescence, Western Blot
OTI11D8
Unconjugated
PASK
Liquid
PASK
Full length human recombit protein of human PASK produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit