Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Patched 1/PTCH Mouse anti-Human, Mouse, Rat, DyLight 755, Clone: OTI1H10, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273254IR
Description
Patched 1/PTCH Monoclonal specifically detects Patched 1/PTCH in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Patched 1/PTCH | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PTCH1 | |
Recombinant protein fragment corresponding to amino acids 770-1027 of human Patched 1/PTCH (NP_000255) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot | |
OTI1H10 | |
Western Blot | |
BCNSFLJ42602, FLJ26746, HPE7, NBCCS, patched (Drosophila) homolog, patched 1, patched homolog (Drosophila), patched homolog 1, patched homolog 1 (Drosophila), protein patched homolog 1, PTC, PTC1, PTCH, PTCH protein +12b, PTCH protein +4′, PTCH protein -10, PTCH11 | |
Mouse | |
Affinity Purified | |
Cell Cycle and Replication, Stem Cell Signaling Pathway | |
5727.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title